Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203713.1 | NP_989044.1 | 1 - 2 | 256 | 102 | |
GenBank mRNA | BC061364.1 | - | 1 - 2 | 256 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |