Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203713.1 | NP_989044.1 | 3 - 4 | 594 | 66 | |
GenBank mRNA | BC061364.1 | - | 3 - 4 | 594 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |