Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam98a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203730.2 | NP_989061.2 | 1 - 2 | 364 | 83 | |
GenBank mRNA | BC061314.1 | - | 1 - 2 | 356 | 83 | |
CX502999.2 | - | 1 - 2 | 364 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |