Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: omp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203734.1 | NP_989065.2 | 1 | 451 | 61 | |
GenBank mRNA | BC061304.1 | - | 1 | 451 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |