Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203737.1 | NP_989068.1 | 3 - 4 | 311 | 72 | |
GenBank mRNA | BC061298.1 | - | 3 - 4 | 311 | 72 | |
BC168799.1 | - | 2 - 3 | 287 | 72 | ||
CR942700.2 | - | 3 - 4 | 301 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |