Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sf3b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203785.2 | NP_989116.1 | 2 - 3 | 200 | 83 | |
GenBank mRNA | BC061357.1 | - | 2 - 3 | 201 | 83 | |
CR760335.2 | - | 2 - 3 | 171 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |