Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp5pb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203787.1 | NP_989118.1 | 5 - 6 | 539 | 118 | |
GenBank mRNA | BC061296.1 | - | 5 - 6 | 539 | 118 | |
CR760878.2 | - | 5 - 6 | 571 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |