Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203792.1 | NP_989123.1 | 6 - 7 | 484 | 78 | |
XM_012958763.1 | XP_012814217.1 | 6 - 7 | 549 | 78 | ||
GenBank mRNA | BC061292.1 | - | 6 - 7 | 484 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |