Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fscn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203797.1 | NP_989128.1 | 4 - 5 | 1288 | 70 | |
XM_018097067.1 | XP_017952556.1 | 4 - 5 | 1406 | 70 | ||
GenBank mRNA | BC061323.1 | - | 4 - 5 | 1288 | 70 | |
CR762320.2 | - | 4 - 5 | 1277 | 70 | ||
DQ374150.1 | - | 4 - 5 | 1284 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |