Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203801.1 | NP_989132.1 | 3 - 4 | 441 | 86 | |
GenBank mRNA | BC061333.1 | - | 3 - 4 | 441 | 86 | |
CR761233.2 | - | 3 - 4 | 440 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |