Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203817.1 | NP_989148.1 | 2 - 3 | 216 | 94 | |
GenBank mRNA | BC061604.1 | - | 2 - 3 | 216 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |