Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plekhj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203835.1 | NP_989166.1 | 5 - 6 | 572 | 88 | |
GenBank mRNA | BC063351.1 | - | 5 - 6 | 572 | 88 | |
CR761359.2 | - | 5 - 6 | 575 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |