Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203853.1 | NP_989184.1 | 4 - 5 | 510 | 98 | |
XM_012964435.2 | XP_012819889.1 | 4 - 5 | 576 | 98 | ||
GenBank mRNA | BC063209.1 | - | 4 - 5 | 510 | 98 | |
CR760309.2 | - | 4 - 5 | 526 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |