Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nucks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203856.2 | NP_989187.1 | 6 - 7 | 1056 | 93 | |
XM_012958321.1 | XP_012813775.1 | 6 - 7 | 1204 | 93 | ||
GenBank mRNA | BC063335.1 | - | 6 - 7 | 782 | 93 | |
CR760325.2 | - | 6 - 7 | 534 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |