Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tktl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203896.1 | NP_989227.1 | 12 - 13 | 1631 | 98 | |
GenBank mRNA | BC063226.1 | - | 12 - 13 | 1631 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |