Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhcr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203904.1 | NP_989235.1 | 134 | 81 | ||
GenBank mRNA | BC063347.1 | - | 134 | 81 | ||
CU075777.1 | - | 110 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |