Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kctd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203908.1 | NP_989239.1 | 3 - 4 | 544 | 92 | |
XM_012960765.2 | XP_012816219.1 | 3 - 4 | 3870 | 92 | ||
XM_018092973.1 | XP_017948462.1 | 3 - 4 | 814 | 92 | ||
GenBank mRNA | BC063913.1 | - | 3 - 4 | 544 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |