Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203933.1 | NP_989264.1 | 2 - 3 | 407 | 71 | |
GenBank mRNA | BC064247.1 | - | 2 - 3 | 407 | 71 | |
BC080335.1 | - | 2 - 3 | 420 | 71 | ||
CR760236.2 | - | 2 - 3 | 358 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |