Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: copz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203948.1 | NP_989279.1 | 6 - 7 | 410 | 66 | |
XM_012956670.2 | XP_012812124.1 | 7 - 8 | 517 | 66 | ||
XM_012956671.2 | XP_012812125.1 | 6 - 7 | 478 | 66 | ||
XM_012956672.2 | XP_012812126.1 | 7 - 8 | 457 | 66 | ||
GenBank mRNA | BC064149.1 | - | 6 - 7 | 410 | 66 | |
CR760836.2 | - | 6 - 7 | 394 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |