Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203969.1 | NP_989300.1 | 3 - 4 | 612 | 78 | |
GenBank mRNA | BC063917.1 | - | 3 - 4 | 612 | 78 | |
BC076710.1 | - | 3 - 4 | 646 | 78 | ||
CT030284.1 | - | 3 - 4 | 588 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |