Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tprkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203994.1 | NP_989325.1 | 2 - 3 | 256 | 113 | |
GenBank mRNA | BC064176.1 | - | 2 - 3 | 256 | 113 | |
CT025420.2 | - | 2 - 3 | 301 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |