Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tprkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203994.1 | NP_989325.1 | 4 - 5 | 554 | 89 | |
GenBank mRNA | BC064176.1 | - | 4 - 5 | 554 | 89 | |
CT025420.2 | - | 4 - 5 | 599 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |