Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4ebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204002.1 | NP_989333.1 | 1 - 2 | 331 | 103 | |
GenBank mRNA | BC064150.1 | - | 1 - 2 | 331 | 103 | |
CT027966.2 | - | 1 - 2 | 167 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |