Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tagln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204023.1 | NP_989354.1 | 2 - 3 | 224 | 100 | |
XM_012969825.1 | XP_012825279.1 | 2 - 3 | 340 | 100 | ||
XM_012969826.1 | XP_012825280.1 | 2 - 3 | 259 | 100 | ||
GenBank mRNA | BC064157.1 | - | 2 - 3 | 224 | 100 | |
CT025168.2 | - | 1 - 2 | 214 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |