Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204028.1 | NP_989359.1 | 6 - 7 | 790 | 78 | |
GenBank mRNA | BC064151.1 | - | 6 - 7 | 790 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |