Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204046.1 | NP_989377.1 | 17 - 18 | 2269 | 70 | |
XM_012971610.2 | XP_012827064.1 | 17 - 18 | 2345 | 70 | ||
GenBank mRNA | BC064723.1 | - | 17 - 18 | 2269 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |