Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204046.1 | NP_989377.1 | 4 - 5 | 608 | 77 | |
XM_012971610.2 | XP_012827064.1 | 4 - 5 | 663 | 77 | ||
GenBank mRNA | BC064723.1 | - | 4 - 5 | 608 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |