Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204079.1 | NP_989410.1 | 2 - 3 | 148 | 80 | |
XM_012958705.2 | XP_012814159.1 | 2 - 3 | 258 | 80 | ||
XM_018091893.1 | XP_017947382.1 | 2 - 3 | 258 | 80 | ||
GenBank mRNA | BC064875.1 | - | 2 - 3 | 148 | 80 | |
BC076991.1 | - | 2 - 3 | 196 | 80 | ||
CR759982.1 | - | 2 - 3 | 127 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |