Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213679.2 | NP_998844.1 | 3 - 4 | 843 | 101 | |
GenBank mRNA | BC067963.2 | - | 3 - 4 | 843 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |