Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012954750.2 | XP_012810204.1 | 1325 | 71 | ||
GenBank mRNA | BC067971.1 | - | 1325 | 71 | ||
BC076953.1 | - | 1311 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |