Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012954750.2 | XP_012810204.1 | 1393 | 105 | ||
GenBank mRNA | BC067971.1 | - | 1393 | 105 | ||
BC076953.1 | - | 1379 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |