Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213698.1 | NP_998863.1 | 9 - 10 | 1379 | 108 | |
XM_012962860.2 | XP_012818314.1 | 9 - 10 | 1354 | 108 | ||
GenBank mRNA | BC067913.1 | - | 9 - 10 | 1379 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |