Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC067328.1 | - | 2 - 3 | 420 | 99 | |
BC080356.1 | - | 2 - 3 | 346 | 99 | ||
CR760270.2 | - | 1 - 2 | 313 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |