Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005126.2 | NP_001005126.1 | 5 - 6 | 303 | 79 | |
XM_018097710.1 | XP_017953199.1 | 5 - 6 | 312 | 79 | ||
XM_018097711.1 | XP_017953200.1 | 5 - 6 | 503 | 79 | ||
GenBank mRNA | BC077662.1 | - | 5 - 6 | 304 | 79 | |
CR848394.2 | - | 4 - 5 | 269 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |