Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrnp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005142.1 | NP_001005142.1 | 4 - 5 | 461 | 75 | |
GenBank mRNA | BC077690.1 | - | 4 - 5 | 461 | 75 | |
CT025161.2 | - | 4 - 5 | 359 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |