Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005144.1 | NP_001005144.1 | 5 - 6 | 513 | 64 | |
GenBank mRNA | BC077693.1 | - | 5 - 6 | 513 | 64 | |
CR761697.2 | - | 5 - 6 | 518 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |