Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005438.1 | NP_001005438.1 | 4 - 5 | 1153 | 102 | |
GenBank mRNA | BC074538.1 | - | 4 - 5 | 1153 | 102 | |
CR761513.2 | - | 4 - 5 | 1157 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Xenopus tropicalis Probe spans exons |