Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dmap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005440.1 | NP_001005440.1 | 10 - 11 | 1376 | 92 | |
GenBank mRNA | BC074546.1 | - | 10 - 11 | 1376 | 92 | |
CR760331.2 | - | 10 - 11 | 1354 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |