Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mkrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005447.1 | NP_001005447.1 | 258 | 79 | ||
GenBank mRNA | BC074559.1 | - | 258 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |