Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc5g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005622.1 | NP_001005622.1 | 151 | 85 | ||
GenBank mRNA | BC074594.1 | - | 151 | 85 | ||
CR942355.1 | - | 231 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |