Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nt5m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005673.2 | NP_001005673.2 | 1 - 2 | 554 | 110 | |
XM_012970294.2 | XP_012825748.1 | 2 - 3 | 715 | 110 | ||
GenBank mRNA | BC074711.1 | - | 1 - 2 | 554 | 110 | |
EG653972.1 | - | 1 - 2 | 177 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |