Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005690.1 | NP_001005690.1 | 3 - 4 | 441 | 81 | |
XM_012959253.1 | XP_012814707.1 | 4 - 5 | 513 | 81 | ||
XM_012959260.2 | XP_012814714.1 | 5 - 6 | 903 | 81 | ||
GenBank mRNA | BC075116.1 | - | 3 - 4 | 441 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |