Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ethe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005706.2 | NP_001005706.1 | 2 - 3 | 270 | 95 | |
GenBank mRNA | BC075280.1 | - | 2 - 3 | 272 | 95 | |
CR942545.2 | - | 2 - 3 | 267 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |