Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ethe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005706.2 | NP_001005706.1 | 3 - 4 | 424 | 81 | |
GenBank mRNA | BC075280.1 | - | 3 - 4 | 426 | 81 | |
CR942545.2 | - | 3 - 4 | 421 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |