Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005707.1 | NP_001005707.1 | 4 - 5 | 420 | 67 | |
GenBank mRNA | BC075283.1 | - | 4 - 5 | 420 | 67 | |
CT027875.2 | - | 4 - 5 | 343 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |