Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005715.1 | NP_001005715.1 | 2 - 3 | 376 | 86 | |
XM_018092991.1 | XP_017948480.1 | 3 - 4 | 432 | 86 | ||
GenBank mRNA | BC075304.1 | - | 2 - 3 | 376 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |