Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005715.1 | NP_001005715.1 | 3 - 4 | 508 | 104 | |
XM_018092991.1 | XP_017948480.1 | 4 - 5 | 564 | 104 | ||
GenBank mRNA | BC075304.1 | - | 3 - 4 | 508 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |