Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nipsnap3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005718.1 | NP_001005718.1 | 2 - 3 | 419 | 79 | |
XM_012959387.2 | XP_012814841.1 | 2 - 3 | 555 | 79 | ||
GenBank mRNA | BC075307.1 | - | 2 - 3 | 419 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |