Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rwdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005815.1 | NP_001005815.1 | 2 - 3 | 218 | 101 | |
XM_012962878.2 | XP_012818332.1 | 2 - 3 | 153 | 101 | ||
GenBank mRNA | BC075365.1 | - | 2 - 3 | 218 | 101 | |
CR761667.2 | - | 2 - 3 | 190 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |