Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006693.2 | NP_001006694.1 | 8 - 9 | 1058 | 96 | |
NM_001316901.1 | NP_001303830.1 | 8 - 9 | 1058 | 96 | ||
GenBank mRNA | BC075396.1 | - | 8 - 9 | 947 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |